Learn More
Description
- ELISA kit is designed for the quantitative determination of IgA in chicken plasma or serum samples.
- Detection Method: Colorimetric
- Assay Type: Quantitative
- Calibration Range: 12.5 to 400 ng/mL
- Limit of Detection: 2.718 ng/mL
- Sample Volume: 100 μL
- Reactivity: Chicken
- Host Species: Goat
- Label: HRP-conjugate
- Applications: ELISA, Quantification
- Research Area: Veterinary Research, Immunology and Microbiology
- Storage: 4 °C
Specifications
Specifications
| Calibration Range | 12.5 to 400ng/mL |
| Conjugate | HRP |
| Cross Reactivity | Chicken |
| Description | IgA (Chicken) ELISA Kit is an enzyme immunoassay for the quantitative determination of IgA in chicken biological samples. |
| Detection Method | Colorimetric |
| Includes | 50mL Diluent concentrate, 50mL wash solution concentrate, 150μL Enzyme Antibody Conjugate concentrate, 12mL TMB Substrate solution, 12mL stop solution, 96-well microtiter plate, 1 vial calibrator (lyophilized) |
| Quantity | 1 Kit |
| Sample Type | Biological Sample |
| Sample Volume | 100 μL |
| Content And Storage | Store the kit at 4°C |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.