Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ IgA (Human) ELISA Kit

Catalog No. 89100368 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
Cell Culture Samples, CSF, Milk, Plasma, Saliva, Serum, Urine
Serum, Plasma
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-100-368 Cell Culture Samples, CSF, Milk, Plasma, Saliva, Serum, Urine
89-100-351 Serum, Plasma
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-368 Supplier Abnova™ Supplier No. KA2110
Add to Cart
Add to Cart

Includes

  • Human IgA Microplate: A 96-well polystyrene microplate coated with a polyclonal antibody against human IgA
  • Sealing Tapes
  • Human IgA Standard: Human IgA in a buffered protein base, lyophilized, 100ng
  • Biotinylated Human IgA Antibody (60x): A 60-fold concentrated biotinylated polyclonal antibody against IgA
  • MIX Diluent Concentrate (10x)
  • Wash Buffer Concentrate (20x)
  • Streptavidin-Peroxidase Conjugate (SP Conjugate): A 100-fold concentrate
  • Chromogen Substrate: A ready-to-use stabilized peroxidase chromogen substrate tetramethylbenzidine
  • Stop Solution: A 0.5N hydrochloric acid

This assay employs a quantitative sandwich enzyme immunoassay technique that measures human IgA in less than 4 hours

Specifications

Calibration Range 0.781 to 50ng/mL
Conjugate Biotin
Cross Reactivity Human
Description IgA (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human IgA.
Detection Limit 0.52ng/mL
Detection Method Colorimetric
Includes Antibody coated microwells (96 wells), Human IgA Standard (200ng), Biotinylated IgA Antibody (50x): 1 vial, MIX Diluent Concentrate (10x): 30mL, Wash Buffer Concentrate (20x): 30mL x 2, Streptavidin-Peroxidase Conjugate (SP Conjugate): 80, Chromogen Substrate (8mL), Stop solution (12mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Cell Culture Samples, CSF, Milk, Plasma, Saliva, Serum, Urine
Sample Volume 50 μL
Packaging 96-well
Content And Storage Store SP conjugate and biotinylated antibody and standard at -20°C. Store all other components at 4°C.
For Use With (Application) Quantification
Product Type ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.