Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ IgA (Rat) ELISA Kit

Catalog No. 89100696 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
Plasma, Serum
Serum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-100-696 Serum
89-028-665 Plasma, Serum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-696 Supplier Abnova™ Supplier No. KA2506
Add to Cart
Add to Cart

Enzyme-linked immunoassay for the quantitative determination of IgA in Rat biological samples

Specifications

Conjugate HRP
Cross Reactivity Rat
Description IgA (Rat) ELISA Kit is for the quantitative measurement of IgA in serum.
Detection Method Colorimetric
Includes Component, Anti-rat IgA coated 96-well plate, Reference standard (Lyophilized ), 10x Immunoglobulin Diluent (25mL), HRP Conjugate Reagent (11mL), 20x Wash Solution (50mL), TMB Reagent (One-step) (11mL), Stop Solution (1N HCl) (11mL)
Quantity 96 Assays
Regulatory Status For research use only (RUO)
Sample Type Serum
Sample Volume 100 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type IgA (Rat) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.