Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ IgE (Human) ELISA Kit, Version 03

Catalog No. 89022695 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-022-695 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-022-695 Supplier Abnova™ Supplier No. KA0216
Add to Cart
Add to Cart

Sandwich enzyme immunoassay for the quantitative measurement of IgE

Specifications

Antigen IgE
Calibration Range 0 to 800IU/mL
Conjugate HRP
Cross Reactivity Human
Description IgE (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of IgE.
Detection Limit 5IU/mL
Detection Method Colorimetric
Includes 96-well Antibody coated well, 13mL Zero Buffer, 18mL Enzyme Conjugate reagent, 1 set reference standard set: 0, 10, 50, 100, 400 and 800 IU/mL lyophilized, 11mL TMB reagent, 11mL stop solution
Quantity 1 Kit
Sample Type Serum
Sample Volume 20 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type IgE (Human) ELISA Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.