Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Igf1 (Mouse), Biotin, ELISA Kit

Catalog No. 89100230 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-230 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-230 Supplier Abnova™ Supplier No. KA0493
Add to Cart
Add to Cart

Includes

  • antibody coated microwells 96 wells
  • Mouse IGF-1 Standard 40ng
  • Biotinylated IGF-1 antibody (50x) 1 vial
  • MIX Diluent Concentrate (10x) 30mL
  • Wash Buffer Concentrate (20x) 30mL x 2
  • Streptavidin-Peroxidase Conjugate (SP Conjugate) 80
  • Chromogen Substrate 8mL
  • Stop Solution: 12mL

Sandwich Enzyme Immunoassay for the quantitative measurement of pig albumin

Specifications

Calibration Range 0.16 to 10ng/mL
Conjugate Biotin
Cross Reactivity Mouse
Description Igf1 (Mouse) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of mouse Igf1.
Detection Limit 0.1 ng/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Cell culture supernatant, plasma and serum
Sample Volume 50 μL
Content And Storage Store components of the kit at 4°C or -20°C as described in the protocol.
For Use With (Application) Quantification
Product Type Igf1 (Mouse) ELISA Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.