Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ IgG (Rat) ELISA Kit

Catalog No. 89028667 Shop All Abnova Corporation Products
Change view
Click to view available options
Product Type:
IgG (Rat) ELISA kit
Calibration Range:
1.95 to 125 ng/mL
25 to 800 ng/mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Calibration Range
89-028-667 IgG (Rat) ELISA kit 25 to 800 ng/mL
89-100-697 IgG (Rat) ELISA kit 1.95 to 125 ng/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-028-667 Supplier Abnova™ Supplier No. KA1952
Add to Cart
Add to Cart

Includes

  • Diluent Concentrate (Running buffer): One bottle containing 5X concentrated diluent running buffer, 50mL
  • Wash Solution Concentrate: One bottle containing 20X concentrated wash solution, 50mL
  • Enzyme-Antibody Conjugate 100X: One vial containing affinity purified anti- Rat IgG antibody conjugated with horseradish peroxidase in a stabilizing buffer, 150μL
  • Chromogen-Substrate Solution: One vial containing 3,3',5,5'-tetramethylbenzidine (TMB) and hydrogen peroxide in citric acid buffer at pH 3.3, 12mL
  • Stop Solution: One vial containing 0.3M sulfuric acid (WARNING: Avoid contact with skin), 12mL
  • Anti-Rat IgG ELISA Micro Plate: Twelve removable eight well strips in well holder frame. Each well is coated with affinity purified anti-Rat IgG, 96 (8 x 12) wells
  • Rat IgG Calibrator: One vial containing Rat IgG calibrator, 1 vial

Enzyme-linked immunoassay for the quantitative determination of IgG in Rat biological samples

Specifications

Calibration Range 25 to 800 ng/mL
Conjugate HRP
Cross Reactivity Rat
Description IgG (Rat) ELISA Kit is a highly sensitive two-site enzyme-linked immunoassay for the quantitative determination of IgG in rat biological samples.
Detection Method Colorimetric
Includes 50mL Diluent concentrate, 50mL wash solution concentrate, 150μL Enzyme Antibody Conjugate concentrate, 12mL TMB Substrate solution, 12mL stop solution, 96-well microtiter plate, 1 vial calibrator (lyophilized)
Quantity 1 Kit
Sample Type Plasma, Serum
Sample Volume 100 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type IgG (Rat) ELISA kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.