Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ IgG1 (Mouse) ELISA Kit

Catalog No. 89100658 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
Plasma, Serum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-100-658 Plasma, Serum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-658 Supplier Abnova™ Supplier No. KA2468
Add to Cart
Add to Cart

Colorimetric immunometric Enzyme Immunoassay kit for the quantitative measurement of Mouse IgG1 in culture supernatants and Serum of Mouse origin

Specifications

Calibration Range 1.56 to 50ng/mL
Conjugate HRP
Cross Reactivity Mouse
Description IgG1 (Mouse) ELISA Kit is for the quantitative measurement of IgG1 in serum.
Detection Method Colorimetric
Includes 96 wells Anti mouse IgG coated wells, Anti mouse IgG1-HRP conjugate (11mL), Reference standard, 20X Wash Solution (50mL), 10X Immunoglobulin Diluent (25mL), TMB Reagent (11mL), Stop Solution (11mL)
Quantity 96 Assays
Regulatory Status For research use only (RUO)
Sample Type Plasma, Serum
Sample Volume 100 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type IgG1 (Mouse) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.