Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ IgG2b (Mouse) ELISA Kit

Catalog No. 89028178 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
Plasma, Serum
Serum, Tissue Culture Medium
Catalog No. Sample Type
89-028-178 Serum, Tissue Culture Medium
89-100-660 Plasma, Serum
2 options

Catalog No. 89-028-178

Supplier: Abnova™ KA1012

Only null left
Add to Cart
Add to Cart

Colorimetric immunometric Enzyme Immunoassay kit for the quantitative measurement of Mouse IgG2b in culture supernatants and Serum of Mouse origin

Specifications

Calibration Range 7.8 to 250ng/mL
Conjugate HRP
Cross Reactivity Mouse
Description IgG2b (Mouse) ELISA Kit is a colorimetric immunometric enzyme immunoassay kit for the quantitative measurement of mouse IgG2b in culture supernatants and serum of mouse origin.
Detection Limit 1.05ng/mL
Detection Method Colorimetric
Includes 96-well microwell, 5mL IgG2b Conjugate, 45mL Assay Buffer concentrate, 100mL Wash Buffer concentrate, 2 standard vials, 10mL TMB Substrate, 10mL stop solution
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Serum, Tissue Culture Medium
Sample Volume 50 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type IgG2b (Mouse) ELISA kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.