Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ IgM (Mouse) ELISA Kit

Catalog No. 89028176 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-176 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-176 Supplier Abnova™ Supplier No. KA1014
Add to Cart
Add to Cart

Colorimetric immunometric Enzyme Immunoassay kit for the quantitative measurement of Mouse IgM in Ascites fluid, culture supernatants and Serum of Mouse origin

Specifications

Calibration Range 3.91 to 250ng/mL
Conjugate HRP
Cross Reactivity Mouse
Description IgM (Mouse) ELISA Kit is a colorimetric immunometric enzyme immunoassay kit for the quantitative measurement of mouse IgM in ascites fluid, culture supernatants, and serum of mouse origin.
Detection Limit 0.6ng/mL
Detection Method Colorimetric
Includes 96-well microwell, 6mL Mouse IgM Conjugate, 50mL Assay Buffer concentrate, 100mL Wash Buffer concentrate, 2 standard vials, 12mL TMB Substrate, 11mL stop solution
Quantity 1 Kit
Sample Type Ascite Fluid, Culture Medium
Sample Volume 50 μL
Content And Storage Store control at -20°C. Store all other components at 4°C.
For Use With (Application) Quantification
Product Type IgM (Mouse) ELISA kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.