Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ IgM (Turkey) ELISA Kit

Catalog No. 89100706 Shop All Abnova Corporation Products
Catalog No. 89-100-706 Supplier Abnova™ Supplier No. KA2516
Add to Cart
Add to Cart

Use to measure quantitatively IgM in plasma, serum and egg-yolk extracts.

IgM (Turkey) ELISA Kit is an enzyme immunoassay for the quantitative determination of IgM in turkey serum, plasma or egg-yolk extracts

Quantification

Specifications

Conjugate HRP
Description IgM (Turkey) ELISA Kit is for the quantitative measurement of IgM in plasma, serum and egg-yolk extracts.
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type IgM (Turkey) ELISA Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.