Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Il10 (Mouse) ELISA Kit

Catalog No. 89100777 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
Body Fluid, CCS, Serum, Tissue Lysates
CCS, Plasma, Serum, Tissue
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-100-777 Body Fluid, CCS, Serum, Tissue Lysates
89-028-154 CCS, Plasma, Serum, Tissue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-777 Supplier Abnova™ Supplier No. KA3070
Add to Cart
Add to Cart

Sandwich Enzyme Immunoassay for the quantitative measurement of Mouse IL-10

Specifications

Calibration Range 15.6 to 1000pg/mL
Conjugate Biotin
Cross Reactivity Mouse
Description Il10 (Mouse) ELISA Kit is intended for the quantitative measurement of mouse Il10.
Detection Limit 1pg/mL
Detection Method Colorimetric
Includes 96-well plate pre-coated with anti-Mouse IL-10 antibody, Lyophilized IL-10 standards (10ng x 2), Sample/Standard diluent buffer (30mL), Biotin conjugated anti-Mouse IL-10 antibody (Concentrated Dilution: 1:100) (130uL), Antibody diluent buffer (12mL), Avidin-Biotin-Peroxidase Complex (ABC) (Concentrated Dilution: 1:100) (130uL), ABC diluent buffer (12mL), TMB substrate (10mL), Stop solution (10mL), Wash buffer (25X) (30mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Body Fluid, CCS, Serum, Tissue Lysates
Sample Volume 100 μL
Content And Storage Store at 4°C for 4 months, or at -20°C for 8 months.
For Use With (Application) Quantification
Product Type Il10 (Mouse) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.