Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ IL12 (Human) ELISA Kit

Catalog No. 89027006 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-027-006 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-027-006 Supplier Abnova™ Supplier No. KA0647

missing translation for 'validMainframeMsg'

Sandwich Enzyme Immunoassay for the quantitative measurement of Human IL-12

Specifications

Calibration Range 7.8 to 500pg/mL
Conjugate HRP
Cross Reactivity Human
Description IL12 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human IL12.
Detection Limit 7pg/mL
Detection Method Colorimetric
Includes 96-well Antibody coated microwell, 2 vials x 1mL 800pg/mL standard (lyophilized), 10mL Biotinylated Antibody, 12mL HRP Conjugated Avidin, 50mL Wash Buffer 20X concentrate, 100μL sample diluent, 8mL TMB-Substrate, 8mL stop solution
Quantity 1 Kit
Sample Type Cell Culture Supernatant, Plasma, Serum, Urine
Sample Volume 50 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type IL12 (Human) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.