Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ IL12b (Mouse) ELISA Kit

Catalog No. 89101060 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-101-060 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-101-060 Supplier Abnova™ Supplier No. KA3412
Add to Cart
Add to Cart

Includes

  • 96-well plate precoated with anti-mouse IL-12(p40) antibody, 96 (8 x 12) wells
  • Lyophilized recombinant mouse IL-12(p40) standard, 10ng x 2
  • Biotinylated anti-mouse IL-12(p40) antibody(dilution 1:100), 130μL
  • Avidin-Biotin-Peroxidase Complex (ABC) (dilution 1:100), 130μL
  • Sample diluent buffer, 30mL
  • Antibody diluent buffer, 12mL
  • ABC diluent buffer, 12mL
  • TMB color developing agent, 10mL
  • TMB stop solution, 10mL

Use to measure quantitatively mouse Il12b. Sandwich enzyme immunoassay.

Il12b (Mouse) ELISA Kit is based on standard sandwich enzyme-linked immune-sorbent assay technology

Specifications

Calibration Range 15.6 to 1000pg/mL
Conjugate Biotin
Cross Reactivity Mouse
Description Il12b (Mouse) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of mouse Il12b.
Detection Limit <10pg/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status RUO
Sample Type Cell Culture Supernatant, Plasma (Citrate, EDTA, Heparin), Serum
Sample Volume 100 μL
Sensitivity <10 pg/mL
Packaging 96-well
Content And Storage Store the kit at -20°C
For Use With (Application) Quantification
Product Type ELISA Kit
Specificity Natural and recombinant mouse IL-12(p40)
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.