Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ IL16 (Human) ELISA Kit

Catalog No. 89100207 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-207 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-207 Supplier Abnova™ Supplier No. KA0520
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • One 96-well Microtiter Plate pre-coated with anti-human IL-16 monoclonal antibody
  • IL-16 Conjugate (Anti-human IL-16 polyclonal antibody conjugated to horseradish peroxidase with preservative)
  • hGH Standard, 12.8ng/vial (Recombinant human IL-16 in a buffered protein base with preservative, lyophilized)
  • Calibrator Diluent I (For serum/plasma testing)
  • Calibrator Diluent II (For cell culture supernatant testing)
  • Wash Buffer (20X)
  • Substrate A (Buffered solution with H2O2)
  • Substrate B (Buffered solution with TMB)
  • Stop Solution (2N Sulphuric Acid (H2SO4). Caution: Caustic Material!)

Use to measure quantitatively human IL16. Sandwich enzyme immunoassay.

This IL-16 enzyme-linked immunosorbent assay (ELISA) applies a technique called a quantitative sandwich immunoassay

Specifications

Calibration Range 20 to 3200pg/mL
Conjugate HRP
Cross Reactivity Human
Description IL16 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human IL16.
Detection Limit 20pg/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Cell Culture Supernatant, Plasma, Serum
Sample Volume 200 μL
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.