Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ IL17A (Human) ELISA Kit

Catalog No. 89028468 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
96 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-468 96 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-468 Supplier Abnova™ Supplier No. KA1732

missing translation for 'validMainframeMsg'

In-Vitro Enzyme-Linked Immunosorbent assay for the quantitative measurement of Human IL-17A

Specifications

Conjugate Biotin
Cross Reactivity Human
Description IL17A (Human) ELISA Kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human IL17A.
Detection Limit 10pg/mL
Detection Method Colorimetric
Includes 96-well Antibody coated microwell, 25mL Wash Buffer 20X concentrate, 2 vials standard, 30mL assay diluent A, 15mL assay diluent B, 2 vials detection Antibody MSP, 8μL HRP-Streptavidian concentrate, 12mL TMB one-step Substrate reagent, 8m
Quantity 96 Assays
Sample Type Cell Culture Supernatant, Plasma, Serum, Urine
Sample Volume 100 μL
Content And Storage Store the kit at -20°C. Standard should be stored at -20°C or -80°C. Avoid repeated freezing and thawing.
For Use With (Application) Quantification
Product Type IL17A (Human) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.