Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ IL18RAP (Human) ELISA Kit

Catalog No. 89100420 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-420 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-420 Supplier Abnova™ Supplier No. KA2140

Includes

  • IL-18R beta Microplate (Item A): coated with anti-human IL-18R beta
  • Wash Buffer Concentrate (20x) (Item B)
  • Standards (Item C): recombinant human IL-18R beta
  • Assay Diluent D (Item K): 5x concentrated buffer. For Standard/Sample (serum/plasma samples/cell culture medium/urine) diluent
  • Assay Diluent B(Item E): 5x concentrated buffer. For detection antibody and HRP- Streptavidin diluent
  • Detection Antibody IL-18R beta (Item F): biotinylated antihuman IL-18R beta
  • HRP-Streptavidin concentrate (Item G): 500x concentrated HRP-conjugated streptavidin
  • TMB One-Step Substrate Reagent (Item H)
  • Stop Solution (Item I): 0.2M sulfuric acid

Use to measure quantitatively human IL18RAP. Enzyme-linked immunosorbent assay.

This assay employs an antibody specific for human IL-18R beta coated on a 96-well plate

Specifications

Conjugate HRP
Cross Reactivity Human
Description IL18RAP (Human) ELISA Kit is an enzyme-linked immunosorbent assay for the quantitative measurement of human IL18RAP.
Detection Limit 12pg/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status RUO
Sample Type Serum, Plasma, Cell Culture Supernatant, Urine
Sample Volume 100 μL
Sensitivity 12 pg/mL
Packaging 96-well
Content And Storage Store the kit at -20°C
For Use With (Application) Quantification
Product Type IL18RAP (Human) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.