Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ IL1B (Mouse) ELISA Kit

Catalog No. 89100765 Shop All Abnova Corporation Products
Change view
Click to view available options
No. of Assays:
96
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Assays
89-100-765 96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-765 Supplier Abnova™ Supplier No. KA3058
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • One 96-well plate pre-coated with anti-mouse IL-1β antibody, 96 (8 x 12) wells
  • Standard (135pg/mL), 0.5mL
  • Standard diluent buffer, 1.5mL
  • Wash buffer (30x), Dilution: 1:30, 20mL
  • Sample diluent buffer, 6mL
  • HRP conjugated anti-mouse IL-1β antibody (RTU), 6mL
  • Stop solution, 6mL
  • TMB substrate A, 6mL
  • TMB substrate B, 6mL
  • Plate sealer, 2 slices
  • Hermetic bag, 1 bag

Use to measure quantitatively mouse Il1b.

This kit is based on standard sandwich enzyme-linked immune-sorbent assay technology

Specifications

Calibration Range 5 to 100pg/mL
Conjugate HRP
Cross Reactivity Mouse
Description Il1b (Mouse) ELISA Kit is intended for the quantitative measurement of mouse Il1b.
Detection Limit 10pg/mL
Detection Method Colorimetric
Includes One 96-well plate pre-coated with anti-mouse IL-1β antibody (96 (8 x 12) wells, Standard (135pg/mL): 0.5mL, Standard diluent buffer (1.5mL), Wash buffer (30x), Dilution: 1:30 (20mL), Sample diluent buffer (6mL), HRP conjugated anti-mouse IL-1β antibody (RTU): 6mL, Stop solution (6mL), TMB substrate A and B (6mL each), Plate sealer (2 slices), Hermetic bag (1)
Regulatory Status For research use only (RUO)
Sample Type Cell Culture Supernatant, Plasma, Serum, Tissue Sample, Urine
Sample Volume 10 μL
Packaging 96-well
Content And Storage Store at 4°C for 6 months.
For Use With (Application) Quantification
Product Type Il1b (Mouse) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.