Learn More
Description
Includes
- Lyophilized recombinant Human IL-1RA standard, 10ng/tube x 2
- One 96-well plate precoated with anti-Human IL-1RA antibody, 1 in no.
- Sample diluent buffer, 30mL
- Biotinylated anti- Human IL-1RA antibody, dilution 1:100, 130μLl
- Antibody diluent buffer, 12mL
- Avidin-Biotin-Peroxidase Complex (ABC), dilution 1:100, 130μL
- ABC diluent buffer, 12mL
- TMB color developing agent, 10mL
- TMB stop solution, 10mL
- The IL1RN (Human) ELISA Kit is a sandwich enzyme immunoassay designed for the quantitative measurement of human IL1RN.
- Assay Type: Quantitative
- Detection Method: Colorimetric
- Label: HRP-conjugated
- Reactivity: Human
- Suitable Samples: Body fluid, cell culture supernatant, serum, and tissue lysate
- Sample Volume: 100 μL
- Calibration Range: 31.2 pg/mL
- Applications: Quantification
- Research Use Only (RUO)
Specifications
Specifications
| Calibration Range | 31.2 to 2000pg/mL |
| Conjugate | HRP |
| Cross Reactivity | Human |
| Description | IL1RN (Human) ELISA Kit is intended for the quantitative measurement of human IL1RN. |
| Detection Method | Colorimetric |
| Quantity | 1 Kit |
| Sample Type | Body Fluid, CCS, Plasma, Serum, Tissue Lysates |
| Sample Volume | 100 μL |
| Content And Storage | Store at 4°C for 4 months, or at -20°C for 8 months. |
| For Use With (Application) | Quantification |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.