Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ IL25 (Mouse) ELISA Kit

Catalog No. 89100467 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-467 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-467 Supplier Abnova™ Supplier No. KA2230
Add to Cart
Add to Cart

Includes

  • Microstrips Coated w/Capture Antibody, 12 x 8 well Microstrips
  • Protein Standard, Lyophilized (10ng), Red
  • Biotinylated Detection Antibody, Lyophilized, Yellow
  • 400x Streptavidin-HRP, 30μL, Blue
  • Wash Buffer (10x), 50mL, Clear
  • Assay Diluent, 50mL, Clear
  • Ready-to-Use Substrate, 12mL, Brown
  • Stop Solution, 12mL, Clear
  • Adhesive Plate Sealers, 4 Sheets

Use to measure quantitatively mouse Il25. Sandwich enzyme immunoassay.

This immunoassay utilizes the quantitative technique of a “Sandwich” Enzyme-Linked Immunosorbent Assay (ELISA) where the target protein (antigen) is bound in a “sandwich” format by the primary capture antibodies coated to each well-bottom and the secondary detection antibodies added subsequently by the investigator

Specifications

Conjugate Biotin
Cross Reactivity Mouse
Description Il25 (Mouse) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of mouse Il25.
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status RUO
Sample Type Cell Lysate, Cell Supernatants, Plasma and Serum
Sample Volume 100 μL
Sensitivity 63 to 4000 pg/mL
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.