Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ IL2RA (Human) ELISA Kit

Catalog No. 89100205 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
CCS, Plasma (EDTA, Heparin), Serum
CCS, Plasma, Serum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-100-205 CCS, Plasma, Serum
89-101-004 CCS, Plasma (EDTA, Heparin), Serum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-205 Supplier Abnova™ Supplier No. KA0522
Add to Cart
Add to Cart

Includes

  • IL-2 sRα MICROTITER PLATE
  • BIOTIN CONJUGATE
  • AVIDIN CONJUGATE
  • IL-2 sRα STANDARD
  • CALIBRATOR DILUENT I
  • CALIBRATOR DILUENT II
  • WASH BUFFER (20X)
  • SUBSTRATE A
  • SUBSTRATE B
  • STOP SOLUTION: Caution: Caustic Material!

Use to measure quantitatively human IL2RA. Sandwich enzyme immunoassay.

This IL2RA ELISA is a ready-to-use 4.5-hour solid phase immunoassay capable of measuring IL-2 sRα levels in serum, plasma, cell culture supernatant, and other biological fluids in the range of 0 to 2000pg/mL

Specifications

Calibration Range 10 to 2000pg/mL
Conjugate HRP
Cross Reactivity Human
Description IL2RA (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human IL2RA.
Detection Limit 10pg/mL
Detection Method Colorimetric
Includes Antibody coated microwells (96 wells), Biotin conjugate (7mL), Avidin conjugate (12mL), IL-2 sRα standard (4000pg/vial x 2), Calibarator diluent I (25mL), Calibrator diluent II (25mL), Wash buffer (20X): 60mL, Substrate A (10mL), Substrate B (10mL), Stop solution (14mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type CCS, Plasma, Serum
Sample Volume 100 μL
Sensitivity 10 pg/mL
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.