Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ IL2RA (Mouse) ELISA Kit

Catalog No. 89101005 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-101-005 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-101-005 Supplier Abnova™ Supplier No. KA3326

missing translation for 'validMainframeMsg'

Includes

  • 96-well plate precoated with anti-mouse CD25 antibody, 96 (8 x 12) wells
  • Lyophilized recombinant mouse CD25 standard, 10ng x 2
  • Biotinylated anti-mouse CD25 antibody (dilution 1:100), 130μL
  • Avidin-Biotin-Peroxidase Complex (ABC) (dilution 1:100), 130μL
  • Sample diluent buffer, 30mL
  • Antibody diluent buffer, 12mL
  • ABC diluent buffer, 12mL
  • TMB color developing agent, 10mL
  • TMB stop solution, 10mL

Use to measure quantitatively mouse Il2ra. Sandwich enzyme immunoassay.

Il2ra (Mouse) ELISA Kit is based on standard sandwich enzyme-linked immune-sorbent assay technology

Specifications

Calibration Range 78 to 5000pg/mL
Conjugate Biotin
Cross Reactivity Mouse
Detection Limit 5pg/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status RUO
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum
Sample Volume 100 μL
Sensitivity <5 pg/mL
Content And Storage Store the kit at -20°C
Product Type ELISA Kit
Specificity Natural and recombinant mouse CD25
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.