Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ IL3 (Human) ELISA Kit

Catalog No. 89100303 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-303 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-303 Supplier Abnova™ Supplier No. KA0360
Add to Cart
Add to Cart

Includes

  • Lyophilized recombinant human IL-3 standard: 10ng/tube x 2
  • One 96-well plate precoated with anti-human IL-3 antibody
  • Sample diluent buffer: 30mL
  • Biotinylated anti-human IL-3 antibody : 130μL, dilution 1:100
  • Antibody diluent buffer: 12mL
  • Avidin-Biotin-Peroxidase Complex (ABC) : 130μL, dilution 1:100
  • ABC diluent buffer: 12mL
  • TMB color developing agent: 10mL
  • TMB stop solution: 10mL

Use to measure quantitatively human IL3. Sandwich enzyme immunoassay.

The human IL-3 ELISA Kit is based on standard sandwich enzyme-linked immune-sorbent assay technology

Specifications

Calibration Range 15.6 to 1000pg/mL
Conjugate Biotin
Cross Reactivity Human
Description IL3 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human IL3.
Detection Limit 1pg/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Body Fluid, Cell Culture Supernatant, Plasma, Serum, Tissue Lysate
Sample Volume 100 μL
Sensitivity <1 pg/mL
Packaging 96-well
Content And Storage Store the kit at -20°C
For Use With (Application) Quantification
Product Type ELISA Kit
Specificity No detectable cross-reactivity with any other cytokine
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.