Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ IL5 (Mouse) ELISA Kit

Catalog No. 89100772 Shop All Abnova Corporation Products
Catalog No. 89-100-772 Supplier Abnova™ Supplier No. KA3065
Add to Cart
Add to Cart

Includes

  • One 96-well plate pre-coated with anti-Mouse IL-5 antibody, 96 (8 x 12) wells
  • Lyophilized Mouse IL-5 standards, 10ng x 2
  • Sample/Standard diluent buffer, 30mL
  • Biotin conjugated anti-Mouse IL-5 antibody (Concentrated), Dilution: 1:100, 130μL
  • Antibody diluent buffer, 12mL
  • Avidin-Biotin-Peroxidase Complex (ABC) (Concentrated), Dilution: 1:100, 130μL
  • ABC diluent buffer, 12mL
  • TMB substrate, 10mL
  • Stop solution, 10mL
  • Wash buffer (25X), 30mL

Use to measure quantitatively mouse Il5.

This kit is based on sandwich enzyme-linked immune-sorbent assay technology

Specifications

Calibration Range 15.6 to 1000pg/mL
Conjugate Biotin
Cross Reactivity Mouse
Description Il5 (Mouse) ELISA Kit is intended for the quantitative measurement of mouse Il5.
Detection Limit 2pg/mL
Detection Method Colorimetric
Includes One 96-well plate pre-coated with anti-Mouse IL-5 antibody (96 (9 x 12) wells), Lyophilized Lyophilized Mouse IL-5 standards Mouse IL-5 standards (10ng x 2), Sample/Standard diluent buffer (30mL), Biotin conjugated anti-Mouse IL-5 antibody (Concentrated), Dilution: 1:100 (130uL), Antibody diluent buffer (12mL), Avidin-Biotin-Peroxidase Complex (ABC) (Concentrated), Dilution: 1:100 (130uL), ABC diluent buffer (12mL), TMB substrate (10mL), Stop solution (10mL), Wash buffer (25X): 30mL
Regulatory Status For research use only (RUO)
Sample Type Body Fluid, Cell Culture Supernate, Serum, Tissue Lysate
Sample Volume 100 μL
Sensitivity <2 pg/mL
Packaging 96-well
Content And Storage Store at 4°C for 4 months, or at -20°C for 8 months.
For Use With (Application) Quantification
Product Type Il5 (Mouse) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.