Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ IL6 (Human) ELISA Kit

Catalog No. 89100773 Shop All Abnova Corporation Products
Catalog No. 89-100-773 Supplier Abnova™ Supplier No. KA3066
Add to Cart
Add to Cart

Sandwich Enzyme Immunoassay for the quantitative measurement of Human IGFBP1

Specifications

Calibration Range 4.69 to 300pg/mL
Description IL6 (Human) ELISA Kit is intended for the quantitative measurement of human IL6.
Detection Method Colorimetric
Sample Type Body Fluid, Cell Culture Supernate, Plasma, Serum, Tissue Lysate
Sample Volume 100 μL
Content And Storage Store at 4°C for 6 months, or at -20°C for 12 months.
For Use With (Application) Quantification
Product Type IL6 (Human) ELISA Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.