Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Use to measure quantitatively human IL6R. Sandwich enzyme immunoassay.

Catalog No. 89100190 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
CCS, Plasma (EDTA, Heparin), Saliva, Serum, Urine
CCS, Plasma, Serum
Conjugate:
Biotin
HRP
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type Conjugate
89-100-190 CCS, Plasma, Serum HRP
89-101-025 CCS, Plasma (EDTA, Heparin), Saliva, Serum, Urine Biotin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-190 Supplier Abnova™ Supplier No. KA0523
Add to Cart
Add to Cart

Includes

  • IL-6sR Microtiter plate, 96 wells
  • Biotin Conjugate, 7mL
  • Avidin Conjugate, 12mL
  • IL-6sR Standard, 2 vials
  • Calibrator diluent I (5x), 25mL
  • Calibrator diluent II, 25mL
  • Wash Buffer (20x), 60mL
  • Substrate A, 10mL
  • Substrate B, 10mL
  • Stop solution, 14mL

Use to measure quantitatively human IL6R. Sandwich enzyme immunoassay.

This IL-6 sR enzyme linked immunosorbent assay (ELISA) applies a technique called a quantitative sandwich immunoassay

Specifications

Calibration Range 31.2 to 2000pg/mL
Conjugate HRP
Cross Reactivity Human
Description IL6R (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human IL6R.
Detection Limit 31.2pg/mL
Detection Method Colorimetric
Includes Antibody coated microwells (96 wells), BIOTIN CONJUGATE (7mL), AVIDIN CONJUGATE (12mL), IL-6 sR STANDARD 4ng/vial x 2, CALIBRATOR DILUENT I (5X) (25mL), CALIBRATOR DILUENT II (25mL), WASH BUFFER (20X) (60mL), SUBSTRATE A (10mL), SUBSTRATE B (10mL), Stop Solution (14mL)
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type CCS, Plasma, Serum
Sample Volume 100 μL
Sensitivity <7 pg/mL
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.