Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Use to measure quantitatively human IL6R. Sandwich enzyme immunoassay.

Catalog No. 89101025 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
CCS, Plasma (EDTA, Heparin), Saliva, Serum, Urine
CCS, Plasma, Serum
Conjugate:
Biotin
HRP
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type Conjugate
89-101-025 CCS, Plasma (EDTA, Heparin), Saliva, Serum, Urine Biotin
89-100-190 CCS, Plasma, Serum HRP
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-101-025 Supplier Abnova™ Supplier No. KA3346
Add to Cart
Add to Cart

Includes

  • 96-well plate precoated with anti-human IL-6R Alpha antibody, 96 (8 x 12) wells
  • Lyophilized recombinant human IL-6R Alpha standard, 10ng x 2
  • Biotinylated anti-human IL-6R Alpha antibody (dilution 1:100), 130μL
  • Avidin-Biotin-Peroxidase Complex (ABC) (dilution 1:100), 130μL
  • Sample diluent buffer, 30mL
  • Antibody diluent buffer, 12mL
  • ABC diluent buffer, 12mL
  • TMB color developing agent, 10mL
  • TMB stop solution, 10mL

Use to measure quantitatively human IL6R. Sandwich enzyme immunoassay.

IL6R (Human) ELISA Kit was based on standard sandwich enzyme-linked immune-sorbent assay technology

Specifications

Calibration Range 31.2 to 2000pg/mL
Conjugate Biotin
Cross Reactivity Human
Description IL6R (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human IL6R.
Detection Method Colorimetric
Includes 96-well plate precoated with anti- human IL-6R Alpha antibody 96 (8 x 12) wells Lyophilized recombinant human IL-6R Alpha standard 10ng x 2, Biotinylated anti- human IL-6R Alpha antibody(dilution 1:100) 130μL, Avidin-Biotin-Peroxidase Complex (ABC) (dilution 1:100) 130μL, Sample diluent buffer 30mL, Antibody diluent buffer 12mL, ABC diluent buffer 12mL, TMB color developing agent 10mL, TMB stop solution 10mL
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type CCS, Plasma (EDTA, Heparin), Saliva, Serum, Urine
Sample Volume 100 μL
Sensitivity <10 pg/mL
Packaging 96-well
Content And Storage Store the kit at -20°C
For Use With (Application) Quantification
Product Type ELISA Kit
Specificity Natural and recombinant human IL-6R Alpha
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.