Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Il7 (Mouse) ELISA Kit

Catalog No. 89027205 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
Body Fluid, CCS, Serum, Tissue Lysates
Cell Lysates, Plasma, Serum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-027-205 Body Fluid, CCS, Serum, Tissue Lysates
89-100-774 Cell Lysates, Plasma, Serum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-027-205 Supplier Abnova™ Supplier No. KA1067
Add to Cart
Add to Cart

Includes

  • Lyophilized recombinant mouse IL-7 standard, 10ng x 2
  • Plate precoated with anti -mouse IL-7 antibody, 96-wells
  • Sample diluent buffer, 30mL
  • Biotinylated anti-mouse IL-7 antibody, dilution 1:100, 130μL
  • Antibody diluent buffer, 12mL
  • Avidin-Biotin-Peroxidase complex (ABC), dilution 1:100, 130μL
  • ABC diluent buffer, 12mL
  • TMB color developing agent, 10mL
  • TMB stop solution, 10mL

Sandwich Enzyme Immunoassay for the quantitative measurement of Mouse IL-7

Specifications

Calibration Range 15.6 to 1000pg/mL
Conjugate Biotin
Cross Reactivity Human, Mouse
Description Mouse Il7 ELISA kit is a sandwich enzyme immunoassay for the quantitative measurement of mouse Il7.
Detection Limit 1pg/mL
Detection Method Colorimetric
Includes 96-well Antibody coated microwell, 2 vials x 10ng recombinant Mouse Il7 standard (lyophilized), 30mL Sample Diluent Buffer, 130μL Biotinylated Anti-Mouse Il7 Antibody, 12mL Antibody Diluent Buffer,, 130μL Avidin-Biotin-
Quantity 1 Kit
Sample Type Body Fluid, CCS, Serum, Tissue Lysates
Sample Volume 100 μL
Content And Storage Store the kit at -20°C
For Use With (Application) Quantification
Product Type Il7 (Mouse) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.