Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Imidazole ELISA Kit

Catalog No. 89024541 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-541 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-541 Supplier Abnova™ Supplier No. KA1421
Add to Cart
Add to Cart

Competitive binding Enzyme Immunoassay for the quantitative measurement of Imidazole

Specifications

Conjugate AP
Description Imidazole ELISA Kit is a competitive binding enzyme immunoassay for the quantitative measurement of imidazole.
Detection Method Colorimetric
Includes 12 Strip of 8 detachable 96-well microtiter plate, 1.2mL negative calibrator, 3 vials x 0.9mL positive calibrator, 10.5mL Imidazole-Alkaline Phosphatase Conjugate (IDZ-ALP), 10.5mL p-Nitrophenyl Phosphate (pNPP) Substrate, 15mL Wash Buffer,, 5.5mL stop solution
Quantity 1 Kit
Sample Type Water
Sample Volume 50 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type Imidazole ELISA kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.