Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ In Situ H3-K9 Acetylation Assay Kit

Catalog No. 89027044 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-027-044 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-027-044 Supplier Abnova™ Supplier No. KA0687
Add to Cart
Add to Cart

Measuring in-situ Acetylation of Histone H3-K9

Specifications

Conjugate HRP
Description In Situ H3-K9 Acetylation Assay Kit is use for measuring in situ acetylation of histone H3-K9.
Detection Method Colorimetric
Includes 2 x 30mL GH1 (Wash Buffer 10X), 2 x 30mL GH2 (Permeabilizing Buffer), 2 x 20mL GH3 (Blocking Buffer), 2 x 15mL GH4 (Antibody Buffer), 10μL GH5 (capture Antibody, 1mg/mL), 40μL GH6 (detection Antibody, 400μ
Quantity 1 Kit
Content And Storage Store GH6 and acetylated histone H3-K9 control at -20°C. Store GH3, GH5, GH7 and 8 well strips at 4°C away from light. Store all other components at room temperature.
For Use With (Application) Quantification
Product Type In Situ H3-K9 Acetylation Assay Kit
No. of Assays 96
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.