Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ In Situ Histone H3-K27 Tri-Methylation Assay Kit

Catalog No. 89027140 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-027-140 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-027-140 Supplier Abnova™ Supplier No. KA0577
Add to Cart
Add to Cart

Includes

  • GK1 (10X Wash buffer), 30mL
  • GK2 (Permeabilizing buffer), 30mL
  • GK3 (Blocking buffer), 20mL
  • GK4 (Antibody buffer), 10mL
  • GK5 (Capture Antibody, 1000μg/mL), 9μL
  • GK6 (Detection Antibody, 200μg/mL), 10μL
  • GK7 (Developing solution), 12mL
  • GK8 (Stop solution), 6mL
  • 30% H2O2 solution 0.5,mL
  • Tri-methyl H3-K27 control (20μg/mL), 15μL
  • 8 well assay strip, 2
  • Microplate, 1

Measuring in-situ tri-Methylation of Histone H3-K27

Specifications

Conjugate HRP
Description In Situ Histone H3-K27 Tri-Methylation Assay Kit is use for measuring in situ tri-methylation of histone H3-K27.
Detection Method Colorimetric
Includes 30mL GK1 (Wash Buffer 10X), 30mL GK2 (Permeabilizing Buffer), 20mL GK3 (Blocking Buffer), 10mL GK4 (Antibody Buffer), 9μL GK5 (capture Antibody, 1000μg/mL), 10μL GK6 (detection Antibody, 200μ
Quantity 1 Kit
Content And Storage Store GK6 and Tri-methyl H3-K27 control at -20°C Store all other components at 4°C away from light.
For Use With (Application) Quantification
Product Type In Situ Histone H3-K27 Tri-Methylation Assay Kit
No. of Assays 96
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.