Learn More
Specifications
Specifications
| Conjugate | HRP |
| Description | In Situ Histone H3-K4 Methylation Assay Kit is use for measuring in situ histone H3-K4 methylation. |
| Detection Method | Colorimetric |
| Includes | 30mL GA1 (Wash Buffer 10X), 30mL GA2 (Permeabilizing Buffer), 20mL GA3 (Blocking Buffer), 10mL GA4 (Antibody Buffer), 60μL GA5 (capture Antibody, 1000μg/mL), 20μL GA6 (detection Antibody, 200 |
| Quantity | 1 Kit |
| Content And Storage | Store GA6 and methylated H3-K4 control at -20°C. Store GA3, GA5, GA7 and 8 well strips at 4°C away from light. Store all other components at room temperature. |
| For Use With (Application) | Quantification |
| Product Type | In Situ Histone H3-K4 Methylation Assay Kit |
| No. of Assays | 96 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.