Learn More
Specifications
Specifications
| Conjugate | HRP |
| Description | In Situ Histone H3-K9 Methylation Assay Kit is use for measuring in situ histone H3-K9 methylation. |
| Detection Method | Colorimetric |
| Includes | 30mL GB1 (Wash Buffer 10X), 30mL GB2 (Permeabilizing Buffer), 20mL GB3 (Blocking Buffer), 15mL GB4 (Antibody Buffer), 60μL GB5 (capture Antibody, 1000μg/mL), 20μL GB6 (detection Antibody, 200 |
| Quantity | 1 Kit |
| Content And Storage | Store GB6 and methylated H3-K9 control at -20°C. Store GB3, GB5, GB7 and 8 well strips at 4°C away from light. Store all other components at room temperature. |
| For Use With (Application) | Quantification |
| Product Type | In Situ Histone H3-K9 Methylation Assay Kit |
| No. of Assays | 96 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.