Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ INHBA (Mouse) ELISA Kit

Catalog No. 89100724 Shop All Abnova Corporation Products
Change view
Click to view available options
No. of Assays:
96
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Assays
89-100-724 96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-724 Supplier Abnova™ Supplier No. KA3017
Add to Cart
Add to Cart

Includes

  • One 96-well plate pre-coated with anti-Mouse Activin A antibody, 96 (8 x 12) wells
  • Lyophilized Activin A standards, 10ng x 2
  • Sample/Standard diluent buffer, 30mL
  • Biotin conjugated anti-Mouse Activin A antibody (Concentrated), Dilution: 1:100, 130μL
  • Antibody diluent buffer, 12mL
  • Avidin-Biotin-Peroxidase Complex (ABC) (Concentrated), Dilution: 1:100, 130μL
  • ABC diluent buffer, 12mL
  • TMB substrate, 10mL
  • Stop solution, 10mL
  • Wash buffer (25X), 30mL

Use to measure quantitatively mouse Inhba.

This kit is based on sandwich enzyme-linked immune-sorbent assay technology

Specifications

Calibration Range 62.5 to 4000pg/mL
Conjugate Biotin
Cross Reactivity Mouse
Description Inhba (Mouse) ELISA Kit is intended for the quantitative measurement of mouse Inhba.
Detection Limit 10pg/mL
Detection Method Colorimetric
Regulatory Status For research use only (RUO)
Sample Type Body Fluid, Cell Culture Supernate, Serum, Tissue Lysate
Sample Volume 100 μL
Sensitivity <10 pg/mL
Packaging 96-well
Content And Storage Store at 4°C for 6 months, or at -20°C for 12 months.
For Use With (Application) Quantification
Product Type Inhba (Mouse) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.