Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Insulin (Human) ELISA Kit

Catalog No. 89101131 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
96 Assays
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-101-131 1 Kit
89-024-462 96 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-101-131 Supplier Abnova™ Supplier No. KA3810
Add to Cart
Add to Cart

Includes

  • Microwells coated with Insulin MAb 12x8x1
  • Insulin Standard 1:1 vial (Ready to use) 2mL
  • Insulin Standards 2-6: 5 vials (Ready to use) 1mL
  • Insulin Enzyme Conjugate: 1 vial 1mL
  • Assay Diluent: 1 bottle (Ready to use) 14mL
  • TMB Substrate: 1 bottle (Ready to use) 12mL
  • Stop Solution: 1 bottle (Ready to use) 12mL
  • 20X Wash concentrate: 1 bottle 25mL

For quantitative measurement Insulin in Human Serum or Plasma

Specifications

Calibration Range 0 to 140 μIU/mL
Cross Reactivity Human
Description Insulin (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human insulin.
Detection Method Colorimetric
Quantity 1 Kit
Sample Type Body Fluid, CCS, Plasma, Serum, Tissue Lysates
Sample Volume 50 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type Insulin (Human) ELISA kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.