Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Insulin (Mouse) ELISA Kit

Catalog No. 89101133 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-101-133 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-101-133 Supplier Abnova™ Supplier No. KA3812
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • One 96-well plate pre-coated with anti-Mouse Insulin antibody, 96 (8x12) wells
  • Lyophilized insulin standards: 8 uIU/mL, 16 uIU/mL, 32 uIU/mL, 80 uIU/mL, 140 uIU/mL, 5 tubes
  • HRP conjugated anti-Mouse Insulin antibody (RTU), 6mL
  • TMB substrate A, 7mL
  • TMB substrate B, 7mL
  • Stop solution, 7mL
  • Wash buffer, (25X) 30mL

Use to measure quantitatively mouse insulin. Sandwich enzyme immunoassay.

The kit is based on sandwich enzyme-linked immune-sorbent assay technology

Specifications

Calibration Range 0 to 140μIU/mL
Conjugate HRP
Cross Reactivity Mouse
Description Insulin (Mouse) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of mouse insulin.
Detection Limit <5 uIU/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status RUO
Sample Type Body Fluid, Cell Culture Supernatant, Plasma, Serum, Tissue Lysate
Sample Volume 50 μL
Sensitivity <5 uIU/mL
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.