Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Insulin (Rat) ELISA Kit

Catalog No. 89101132 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-101-132 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-101-132 Supplier Abnova™ Supplier No. KA3811
Add to Cart
Add to Cart

Includes

  • One 96-well plate pre-coated with anti-Rat Insulin antibody, 96 (8x12) wells
  • Lyophilized insulin standards: 5 tubes (8 uIU/mL, 16 uIU/mL, 32 uIU/mL, 80 uIU/mL, 140 uIU/mL), 5 tubes
  • HRP conjugated anti-Rat Insulin antibody (RTU), 6mL
  • TMB substrate A, 7mL
  • TMB substrate B, 7mL
  • Stop solution, 7mL
  • Wash buffer (25X), 30mL

Use to measure quantitatively rat insulin. Sandwich enzyme immunoassay.

This kit is based on sandwich enzyme-linked immune-sorbent assay technology

Specifications

Calibration Range 0 to 140μIU/mL
Conjugate HRP
Cross Reactivity Rat
Description Insulin (Rat) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of rat insulin.
Detection Limit <5 uIU/ml
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Body Fluid, Cell Culture Supernatant, Plasma, Serum, Tissue Lysates
Sample Volume 50 μL
Sensitivity <5 uIU/mL
Packaging 96-well
Content And Storage Store the kit at 4°C. Avoid repeated freezing and thawing.
For Use With (Application) Quantification
Product Type ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.