Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Jo-1 Ab ELISA Kit

Catalog No. 89024568 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-024-568 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-024-568 Supplier Abnova™ Supplier No. KA1276
Add to Cart
Add to Cart

indirect solid phase ELISA for the quantitative measurement of IgG class autoantibodies against Jo-1 in Human Serum or Plasma

Specifications

Conjugate HRP
Cross Reactivity Human
Description Jo-1 Ab ELISA Kit is an indirect solid phase enzyme immunoassay for the quantitative measurement of IgG class autoantibodies against Jo-1 in human serum or plasma.
Detection Limit 1U/mL
Detection Method Colorimetric
Includes 6 Vials, 1.5mL combined calibrator with IgG class Anti-Jo-1 Antibodies, 2 vials x 1.5mL Anti-C1q controls in a Serum/Buffer Matrix, 2 vials x 1.5mL Anti-Jo-1 controls in a Serum/Buffer matrix, 1 vial x 20mL Sample Buffer, 1 vial x 15mL Enzyme Conjugate solution, 1 vial x 15mL TMB Substrate solution, 1 vial x 15mL stop solution, one 96-well plate, 1 vial x 20mL wash solution
Quantity 1 Kit
Sample Type Plasma, Serum
Sample Volume 100 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type Jo-1 Ab ELISA kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.