Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Kappa (Human) ELISA Kit

Catalog No. 89028683 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-683 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-683 Supplier Abnova™ Supplier No. KA1968
Add to Cart
Add to Cart

Includes

  • Diluent Concentrate (Running Buffer): One bottle containing 5X concentrated diluent running buffer, 50mL
  • Wash Solution Concentrate: One bottle containing 20X concentrated wash solution 50mL
  • Enzyme-Antibody Conjugate 100X: One vial containing affinity-purified anti-Human kappa antibody conjugated with horseradish peroxidase in stabilizing buffer, 150μL
  • Chromogen-Substrate Solution: One vial containing 3,3',5,5'-tetramethybenzidine (TMB) and hydrogen peroxide in citric acid buffer at pH 3.3, 12mL
  • Stop Solution: One vial containing 0.3M sulfuric acid (WARNING: Avoid contact with skin), 12mL
  • Anti-Human Kappa ELISA Micro Plate: Twelve removable eight-well strips in well holder frame. Each well is coated with affinity-purified anti-Human kappa, 96 ( 8 x 12) wells
  • Human Kappa Calibrator: One vial containing Human Kappa Calibrator, 1 vial

highly sensitive two-site enzyme-linked Immunoassay for the quantitative determination of kappa in Human biological samples

Specifications

Calibration Range 6.25 to 200ng/mL
Conjugate HRP
Cross Reactivity Human
Description Kappa (Human) ELISA Kit is a highly sensitive two-site enzyme-linked immunoassay for the quantitative determination of kappa in human biological samples.
Detection Method Colorimetric
Includes 50mL Diluent concentrate, 50mL wash solution concentrate, 150μL Enzyme-Antibody Conjugate concentrate, 12mL TMB Substrate solution, 12mL stop solution, 96-well microtiter plate, 1 vial calibrator (lyophilized)
Quantity 1 Kit
Sample Type Biological Sample
Sample Volume 100 μL
Content And Storage Store the calibrator at -20°C for long term storage. Store all other components at 4°C.
For Use With (Application) Quantification
Product Type Kappa (Human) ELISA kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.