Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Kitl (Mouse) ELISA Kit

Catalog No. 89028595 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-595 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-595 Supplier Abnova™ Supplier No. KA1844
Add to Cart
Add to Cart

For quantitative measurement of Mouse Kitl

Specifications

Conjugate Biotin
Cross Reactivity Mouse
Description Kitl (Mouse) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of mouse Kitl.
Detection Limit 0.06ng/mL
Detection Method Colorimetric
Includes 96-well Antibody coated microwell, 4ng Mouse SCF standard, 1 vial 50X Biotinylated SCF Antibody, 30mL 10X MIX Diluent concentrate, 2 x 30mL Wash Buffer 20X concentrate, 80μL Streptavidin-Peroxidase Conjugate (SP Conjugate),, 8mL chromoge
Quantity 1 Kit
Sample Type Cell Culture Supernatant
Sample Volume 50 μL
Content And Storage Store components of the kit at 4°C or -20°C as described in the protocol.
For Use With (Application) Quantification
Product Type Kitl (Mouse) ELISA kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.