Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ KLK1 (Human) ELISA Kit

Catalog No. 89028741 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-028-741 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-028-741 Supplier Abnova™ Supplier No. KA2009
Add to Cart
Add to Cart

Sandwich Enzyme Immunoassay for the quantitative measurement of Human KLK1

Specifications

Calibration Range 156 to 10000pg/mL
Conjugate Biotin
Cross Reactivity Human
Description KLK1 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human KLK1.
Detection Limit <10pg/mL
Detection Method Colorimetric
Includes 96-well Antibody coated microwell, 2 vials x 10ng recombinant Human KLK1 standard (lyophilized), 30mL Sample Diluent Buffer, 130μL Biotinylated Anti-Human KLK1 Antibody, 12mL Antibody Diluent Buffer,, 130μL Avidin-Bioti
Quantity 1 Kit
Sample Type Cell Culture Supernatant, Cell Lysate, Plasma (EDTA, Heparin), Serum, Tissue Lysate
Sample Volume 100 μL
Content And Storage Store the kit at -20°C
For Use With (Application) Quantification
Product Type KLK1 (Human) ELISA kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.