Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ KLK5 (Human) ELISA Kit

Catalog No. 89100513 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-513 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-513 Supplier Abnova™ Supplier No. KA2352
Add to Cart
Add to Cart

Includes

  • 96-well plate precoated with anti-human KLK5 antibody, 96 wells
  • Lyophilized recombinant human KLK5 standard, 10ng/tube x 2
  • Biotinylated anti-human KLK5 antibody, dilution 1:100, 130μL
  • Avidin-Biotin-Peroxidase Complex (ABC), dilution 1:100, 130μL
  • Sample diluent buffer, 30mL
  • Antibody diluent buffer, 12mL
  • ABC diluent buffer, 12mL
  • TMB color developing agent, 10mL
  • TMB stop solution, 10mL

Use to detect quantitatively of human KLK5 in serum, plasma, body fluids, tissue lysates or cell culture supernatant.

KLK5 (Human) ELISA Kit is based on standard sandwich enzyme-linked immune-sorbent assay technology

Specifications

Calibration Range 78 to 5000pg/mL
Conjugate Biotin
Cross Reactivity Human
Description KLK5 (Human) ELISA Kit is an immunoassay for quantitative detection of human KLK5 in serum, plasma, body fluids, tissue lysates or cell culture supernatant.
Detection Limit <10pg/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Cell culture supernatant, Human milk, Plasma (Heparin, EDTA), Saliva, Serum
Sample Volume 100 μL
Sensitivity <10 pg/mL
Packaging 96-well
Content And Storage Store the kit at -20°C
For Use With (Application) Quantification
Product Type ELISA Kit
Specificity Natural and recombinant human KLK5
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.