Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ KLKB1 (Human) ELISA Kit

Catalog No. 89101126 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-101-126 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-101-126 Supplier Abnova™ Supplier No. KA3830
Add to Cart
Add to Cart

Includes

  • Human Prekallikrein Microplate
  • Sealing Tapes: Precut, pressure sensitive sealing tapes that can be cut to fit the format of the individual assay
  • Human Prekallikrein Standard: Human PK in a buffered protein base (lyophilized), 320ng
  • Biotinylated Human Prekallikrein Antibody (50x): A 50-fold concentrated biotinylated polyclonal antibody against human PK
  • MIX Diluent Concentrate (10x)
  • Wash Buffer Concentrate (20x)
  • Streptavidin-Peroxidase Conjugate (SP Conjugate): A 100-fold concentrate
  • Chromogen Substrate: A ready-to-use stabilized peroxidase chromogen substrate tetramethylbenzidine
  • Stop Solution: A 0.5N hydrochloric acid

Use to measure quantitatively human KLKB1. Sandwich enzyme immunoassay.

This assay employs a quantitative sandwich enzyme immunoassay technique that measures PK in less than 4 hours

Specifications

Calibration Range 1.25 to 80.0ng/mL
Conjugate Biotin
Cross Reactivity Human
Description KLKB1 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human KLKB1.
Detection Limit 1.2ng/mL
Detection Method Colorimetric
Quantity 1 Kit
Sample Type Cell Culture Supernatant, CSF, Milk, Plasma, Saliva, Serum
Sample Volume 50 μL
Packaging 96-well
Content And Storage Store the kit at 4°C. For long term storage, please store at -20°C.
For Use With (Application) Quantification
Product Type ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.