Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Kng1l1 (Rat) ELISA Kit

Catalog No. 89100399 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-399 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-399 Supplier Abnova™ Supplier No. KA2139
Add to Cart
Add to Cart

Includes

  • Diluent Concentrate (Running Buffer)
  • Wash Solution Concentrate
  • Enzyme-Antibody Conjugate 100X: 150 μl of affinity purified anti-Rat THIOSTATIN antibody conjugated with horseradish peroxidase in a stabilizing buffer
  • Chromogen-Substrate Solution
  • Stop Solution
  • Anti-Rat Thiostatin Elisa Micro Plate: Twelve removable eight (8) well micro well strips in well holder frame
  • Each well is coated with affinity purified anti-Rat Thiostatin
  • Rat Thiostatin Calibrator: Rat Thiostatin calibrator. For In Vitro Use Only

Measure Rat Kng1l1. Two-site enzyme linked immunoassay.

In this assay the THIOSTATIN present in samples reacts with the anti-THIOSTATIN antibodies which have been adsorbed to the surface of polystyrene microtitre wells

Specifications

Conjugate HRP
Cross Reactivity Rat
Description Kng1l1 (Rat) ELISA Kit is a two-site enzyme linked immunoassay for the measurement of Rat Kng1l1.
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Biological Fluid
Sample Volume 100 μL
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.