Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ LBP (Human) ELISA Kit

Catalog No. 89100257 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-257 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-257 Supplier Abnova™ Supplier No. KA0448
Add to Cart
Add to Cart

Includes

  • Precoated ELISA modules, 96 wells
  • Detecting antibody (POD-labelled monoclonal antibody to human LBP, 10ng/mL) Ready for use, 1 vial
  • Human LBP-standard (2.67μg/mL), 1 vial
  • Reference serum (9.5μg/mL), 1 vial
  • PBS, 2 tablets
  • Dilution Buffer, 1 vial
  • Tween 20, 1 vial
  • Stopping solution Ready for use, 1 vial
  • Substrate solution Ready for use, 1 vial

Use to measure quantitatively human LBP. Sandwich enzyme immunoassay.

The LBP (Human) ELISA Kit is a solid phase sandwich Enzyme Linked-Immunosorbent Assay (ELISA)

Specifications

Calibration Range 1.5 to 50ng/mL
Conjugate POD
Cross Reactivity Human
Description LBP (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human LBP.
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Culture Medium, Plasma, Serum
Sample Volume 100 μL
Packaging 96-well
Content And Storage Store the kit at -20°C. For best results, please store the components as described in the protocol. Avoid repeated freezing and thawing.
For Use With (Application) Quantification
Product Type ELISA Kit
Specificity Free LBP
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.