Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Lbp (Mouse) ELISA Kit

Catalog No. 89022876 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-022-876 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-022-876 Supplier Abnova™ Supplier No. KA0449
Add to Cart
Add to Cart

Sandwich Enzyme Immunoassay for the quantitative measurement of Mouse Lbp

Specifications

Calibration Range 1.56 to 50ng/mL
Conjugate POD
Cross Reactivity Mouse
Description Lbp (Mouse) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of mouse Lbp.
Detection Method Colorimetric
Includes 96-well microwell Precoated, 1 vial detecting Antibody, 1 vial Mouse-LBP-standard, 1 vial reference Serum, 2 tab PBS, 1 vial dilution Buffer, 1 vial Tween-20, 1 vial stop solution, 1 vial Substrate solution
Quantity 1 Kit
Sample Type Culture Medium, Plasma, Serum
Sample Volume 100 μL
Content And Storage Store Mouse LBP-standard and Mouse Reference serum at -20°C or -80°C. Store all other components at 4°C.
For Use With (Application) Quantification
Product Type Lbp (Mouse) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.