Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Lcn2 (Mouse) ELISA Kit

Catalog No. 89100797 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Volume:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Volume
89-100-797 10 μL
89-027-123 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-797 Supplier Abnova™ Supplier No. KA3094
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • 96-well plate precoated with anti-mouse NGAL antibody, 96 (8 x 12) wells
  • Lyophilized recombinant mouse NGAL standard, 10 ng/tube × 2
  • Biotinylated anti- mouse NGAL antibody, dilution 1:100, 130μL
  • Avidin-Biotin-Peroxidase Complex (ABC), dilution 1:100, 130μL
  • Sample diluent buffer, 30mL
  • Antibody diluent buffer, 12mL
  • ABC diluent buffer, 12mL
  • TMB color developing agent, 10mL
  • TMB stop solution, 10mL

Sandwich Enzyme Immunoassay for the quantitative measurement of Mouse Lcn2

Specifications

Calibration Range 156 to 10,000 pg/mL
Description Lcn2 (Mouse) ELISA Kit is intended for the quantitative measurement of mouse Lcn2.
Detection Method Colorimetric
Quantity 1 Kit
Sample Type CCS, Plasma, Serum, Tissue Sample, Urine
Sample Volume 10 μL
Content And Storage Store at 4°C for 6 months.
For Use With (Application) Quantification
Product Type Lcn2 (Mouse) ELISA Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.