Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Lcn2 (Rat), HRP, ELISA Kit

Catalog No. 89100798 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-798 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-798 Supplier Abnova™ Supplier No. KA3095
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • One 96-well plate pre-coated with anti-rat ngAL antibody 96 (8x12) wells
  • Standard (1350 pg/mL) 0.5mL
  • Standard diluent buffer 1.5mL
  • Wash buffer (30x), Dilution: 1:30 20mL
  • Sample diluent buffer 6mL
  • HRP conjugated anti-ratngAL antibody (RTU) 6mL
  • Stop solution 6mL
  • TMB substrate A 6mL
  • TMB substrate B 6mL
  • Plate sealer 2 slices
  • Hermetic bag 1 bag

For quantitative measurement of Mouse Lcn2.

Specifications

Calibration Range 156 to 10000pg/mL
Conjugate HRP
Cross Reactivity Rat
Description Lcn2 (Rat) ELISA Kit is intended for the quantitative measurement of rat Lcn2.
Detection Limit 10 pg/mL
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Cell culture supernatant, plasma, serum, tissue sample and urine
Sample Volume 10 μL
Content And Storage Store at 4°C for 6 months.
For Use With (Application) Quantification
Product Type Lcn2 (Rat) ELISA Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.