Learn More
Description
Specifications
Specifications
| Conjugate | HRP |
| Cross Reactivity | Human |
| Detection Method | Colorimetric |
| Regulatory Status | For research use only (RUO) |
| Sample Type | Plasma (Heparin) |
| Sample Volume | 100 μL |
| Content And Storage | Store standard, capture antibody, detection antibody and sample diluent additive at −20°C; others at 4°C. |
| Product Type | ELISA Kit |
| No. of Assays | 96 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.