Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ LEP (Human) ELISA Kit

Catalog No. 89100783 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
Body Fluid, CCS, Plasma, Serum, Tissue Lysates
Plasma (Citrate, EDTA, Heparin), Serum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-100-783 Body Fluid, CCS, Plasma, Serum, Tissue Lysates
89-100-007 Plasma (Citrate, EDTA, Heparin), Serum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-783 Supplier Abnova™ Supplier No. KA3080
Add to Cart
Add to Cart

Sandwich Enzyme Immunoassay for the quantitative measurement of Human LEP

Specifications

Calibration Range 62.5 to 4000 pg/mL
Conjugate Biotin
Cross Reactivity Human
Description LEP (Human) ELISA Kit is intended for the quantitative measurement of human LEP.
Detection Limit 5pg/mL
Detection Method Colorimetric
Includes 96-well plate pre-coated with anti-Human Leptin antibody, Lyophilized Leptin standards (10ng x 2), Sample/Standard diluent buffer (30mL), Biotin conjugated anti-Human Leptin antibody (Concentrated), Dilution: 1:100 (130μL), ABC diluent buffer (12mL), TMB substrate (10mL), Stop solution (10mL), Wash buffer (25X) (30mL)
Quantity 96 Assays
Sample Type Body Fluid, CCS, Plasma, Serum, Tissue Lysates
Sample Volume 100 μL
Content And Storage Store at 4°C for 4 months, or at -20°C for 8 months.
For Use With (Application) Quantification
Product Type LEP (Human) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.