Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ LEP (Mouse/Rat) ELISA Kit

Catalog No. 89100006 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-100-006 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-100-006 Supplier Abnova™ Supplier No. KA0026
Add to Cart
Add to Cart

Includes

  • Antibody Coated Microtiter Strips (Ready to use), 96 wells
  • Biotin Labelled Antibody Conc. (10x), 1.3mL
  • Streptavidin-HRP Conjugate (Ready to use), 13mL
  • Master Standard Mouse, 2 vials
  • Master Standard Rat, 2 vials
  • Quality Control Mouse, 2 vials
  • Quality Control Rat, 2 vials
  • Dilution Buffer (Ready to use), 13mL x 2
  • Biotin–Ab Diluent (Ready to use), 13mL
  • Wash Solution Conc. (10x), 100mL
  • Substrate Solution (Ready to use), 13mL
  • Stop Solution (Ready to use), 13mL

Use to measure quantitatively mouse and/or rat leptin in serum, plasma and tissue culture medium. Sandwich enzyme immunoassay.

  • The kit measures leptin in serum and plasma (EDTA, citrate, heparin)
  • Quality Controls are mouse and rat serum based. No human sera are used
  • Standards are recombinant protein based
  • Components of the kit are provided ready to use, concentrated or lyophilized

Specifications

Calibration Range 100 to 4000pg/mL
Conjugate HRP
Cross Reactivity Human, Mouse, Rat
Description Lep (Mouse/Rat) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of mouse and/or rat leptin in serum, plasma.
Detection Limit 30pg/mL (mouse), 50pg/mL (rat)
Detection Method Colorimetric
Quantity 1 Kit
Regulatory Status For research use only (RUO)
Sample Type Plasma (Citrate, EDTA, Heparin), Serum
Sample Volume 100 μL
Sensitivity 30 pg/mL
Total Assay Time 3.5 hr.
Packaging 96-well
Content And Storage Store the kit at 4°C
For Use With (Application) Quantification
Product Type ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.